Skip to Content
Merck
All Photos(1)

Key Documents

276820

Sigma-Aldrich

Phosphorus tribromide solution

1.0 M in methylene chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€66.90
800 ML
€153.00

€66.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€66.90
800 ML
€153.00

About This Item

Linear Formula:
PBr3
CAS Number:
Molecular Weight:
270.69
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

€66.90


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

concentration

1.0 M in methylene chloride

density

1.488 g/mL at 25 °C

SMILES string

BrP(Br)Br

InChI

1S/Br3P/c1-4(2)3

InChI key

IPNPIHIZVLFAFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrey S Mereshchenko et al.
Nature chemistry, 7(7), 562-568 (2015-06-24)
'Roaming' is a new and unusual class of reaction mechanism that has recently been discovered in unimolecular dissociation reactions of isolated molecules in the gas phase. It is characterized by frustrated bond cleavage, after which the two incipient fragments 'roam'

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service