Skip to Content
Merck
All Photos(1)

Key Documents

276146

Sigma-Aldrich

Trihexylsilane

95%

Synonym(s):

NSC 139856

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€272.00

€272.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€272.00

About This Item

Linear Formula:
[CH3(CH2)5]3SiH
CAS Number:
Molecular Weight:
284.60
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

€272.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

reagent type: reductant

refractive index

n20/D 1.448 (lit.)

bp

160.5-161 °C (lit.)

density

0.799 g/mL at 25 °C (lit.)

SMILES string

CCCCCC[SiH](CCCCCC)CCCCCC

InChI

1S/C18H40Si/c1-4-7-10-13-16-19(17-14-11-8-5-2)18-15-12-9-6-3/h19H,4-18H2,1-3H3

InChI key

IBNSYFQZHBBNLR-UHFFFAOYSA-N

Application

Reactant for:
  • Enantioselective Si-H insertion
  • Regio- and stereoselective oxidation into silanols
  • Asymmetric carbenoid insertion of diazoacetates into the silicon-hydrogen bond
  • Hydrosilylation of carbon dioxide

Catalyst for hydrodefluorination leading to the synthesis of hydrocarbons

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service