Skip to Content
Merck
All Photos(1)

Key Documents

27614

Sigma-Aldrich

Methyl benzoate

≥99% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH3
CAS Number:
Molecular Weight:
136.15
Beilstein:
1072099
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay:
≥99% (GC)
Pricing and availability is not currently available.

vapor density

4.68 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99% (GC)

impurities

≤0.2% free acid (as C6H5COOH)
≤0.2% water (Karl Fischer)

ign. residue

≤0.05% (as SO4)

refractive index

n20/D 1.516 (lit.)

bp

198-199 °C (lit.)

mp

−12 °C (lit.)

solubility

H2O: soluble 2.1 g/L at 20 °C

density

1.088 g/mL at 20 °C (lit.)

SMILES string

COC(=O)c1ccccc1

InChI

1S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

QPJVMBTYPHYUOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl benzoate is the most abundant volatile scent compound present in snapdragon flowers[1].

Application

Methyl benzoate has been used in benzoylations of both methyl ketone groups of bisacetylferrocene to form the bis-β-diketone[2].

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - Pensky-Martens closed cup

Flash Point(C)

77 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Benzoylations of Both Methyl Ketone Groups of Bisacetylferrocene with Methyl Benzoate and Alkali Amides to Form the Bis-?-diketone. Certain Derivatives1.
Hauser CR and Cain CE.
The Journal of Organic Chemistry, 23(8), 1142-1146 (1958)
N Dudareva et al.
The Plant cell, 12(6), 949-961 (2000-06-15)
In snapdragon flowers, the volatile ester methyl benzoate is the most abundant scent compound. It is synthesized by and emitted from only the upper and lower lobes of petals, where pollinators (bumblebees) come in contact with the flower. Emission of
María Manchado-Rojo et al.
The Plant journal : for cell and molecular biology, 72(2), 294-307 (2012-06-20)
The transcriptional network topology of B function in Antirrhinum, required for petal and stamen development, is thought to rely on initial activation of transcription of DEFICIENS (DEF) and GLOBOSA (GLO), followed by a positive autoregulatory loop maintaining gene expression levels.
Gianfranco Cainelli et al.
Chemistry & biodiversity, 5(5), 811-829 (2008-05-22)
Novel 4-alkylidene-beta-lactams with a polyphenolic side chain were synthesized and evaluated as radical scavengers. We have undertaken a detailed study of the antioxidant activity in vitro with chemical and biological testing of the new beta-lactams and of the corresponding methyl
Nan Zhao et al.
Plant physiology and biochemistry : PPB, 48(4), 279-287 (2010-03-05)
Two benzenoid esters, methyl salicylate (MeSA) and methyl benzoate (MeBA), were detected from insect-damaged rice plants. By correlating metabolite production with gene expression analysis, five candidate genes encoding putative carboxyl methyltransferases were identified. Enzymatic assays with Escherichia coli-expressed recombinant proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service