Skip to Content
Merck
All Photos(1)

Key Documents

275727

Sigma-Aldrich

5-Methoxypsoralen

99%

Synonym(s):

4-Methoxyfuro[3,2-g]benzopyrane-7-one, Bergapten, Geralen, Heraclin, Majudin

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€109.00
1 G
€345.00

€109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€109.00
1 G
€345.00

About This Item

Empirical Formula (Hill Notation):
C12H8O4
CAS Number:
Molecular Weight:
216.19
Beilstein:
19560
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€109.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

mp

190-193 °C (lit.)

SMILES string

COc1c2C=CC(=O)Oc2cc3occc13

InChI

1S/C12H8O4/c1-14-12-7-2-3-11(13)16-10(7)6-9-8(12)4-5-15-9/h2-6H,1H3

InChI key

BGEBZHIAGXMEMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Methoxypsoralen has been studied for its use in oral photochemotherapy in the treatment of psoriasis and other skin diseases.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silke B Bodendiek et al.
European journal of medicinal chemistry, 44(5), 1838-1852 (2008-12-06)
The voltage-gated potassium channel Kv1.3 constitutes an attractive pharmacological target for the treatment of effector memory T cell-mediated autoimmune diseases such as multiple sclerosis and psoriasis. Using 5-methoxypsoralen (5-MOP, 1), a compound isolated from Ruta graveolens, as a template we
M T Conconi et al.
Pharmacology & toxicology, 82(4), 193-198 (1998-05-19)
The in vitro antiproliferative activity and in vivo phototoxicity of some methyl derivatives of 5-methoxypsoralen and 5-methoxyangelicin, i.e. 4,4'-dimethyl-5-methoxyangelicin (compound I), 3,4'-dimethyl-5-methoxyangelicin (compound II), 4,4'-dimethyl-5-methoxypsoralen (compound III); and 3,4'-dimethyl-5-methoxypsoralen (compound IV), have been investigated. The effects of the compounds were
Wei Yang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(5-6), 575-582 (2010-02-02)
A sensitive, specific and rapid liquid chromatography-mass spectrometry (LC-MS) method has been developed and validated for the simultaneous determination of xanthotoxin (8-methoxypsoralen), psoralen, isoimpinellin (5,8-dimethoxypsoralen) and bergapten (5-methoxypsoralen) in rat plasma using pimpinellin as an internal standard (IS). The plasma
Shinsuke Marumoto et al.
Journal of agricultural and food chemistry, 58(13), 7777-7781 (2010-06-10)
The biotransformation of bergapten (1) by the fungus Glomerella cingulata gave the corresponding reduced acid, 6,7-furano-5-methoxy hydrocoumaric acid (2), a new compound. Xanthotoxin (3) was also converted to the corresponding reduced acid cnidiol b (4) and demethylated metabolite xanthotoxol (5)
Jarogniew J Łuszczki et al.
Pharmacological reports : PR, 62(6), 1231-1236 (2011-01-29)
The aim of this study was to determine and compare the anticonvulsant activities of four natural furanocoumarins [bergapten (5-methoxypsoralen), imperatorin (8-isopentenyloxypsoralen), oxypeucedanin (5-epoxy-isopentenyloxypsoralen) and xanthotoxin (8-methoxypsoralen)] in the maximal electroshock-induced seizure test in mice. The anticonvulsant effects of bergapten, imperatorin

Related Content

Nancy-520 for DNA Detection and Quantitation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service