Skip to Content
Merck
All Photos(1)

Key Documents

275190

Sigma-Aldrich

3,3,3-Tris(4-chlorophenyl)propionic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4)3CCH2CO2H
CAS Number:
Molecular Weight:
405.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

190-193 °C (lit.)

SMILES string

OC(=O)CC(c1ccc(Cl)cc1)(c2ccc(Cl)cc2)c3ccc(Cl)cc3

InChI

1S/C21H15Cl3O2/c22-17-7-1-14(2-8-17)21(13-20(25)26,15-3-9-18(23)10-4-15)16-5-11-19(24)12-6-16/h1-12H,13H2,(H,25,26)

InChI key

LHIVWYJOCNGZRI-UHFFFAOYSA-N

Gene Information

human ... MIF(4282)

Application

3,3,3-Tris(4-chlorophenyl)propionic acid was used as a potent inhibitor for Yersinia PTP(protein-tyrosine phosphatase) YopH[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Zhang et al.
Journal of the American Chemical Society, 140(40), 12724-12727 (2018-09-25)
Mechanical bonds are known to efficiently absorb mechanical energy at low forces, but their behavior at high forces is unknown. Here we investigate the impact of a mechanical bond on the rate of activation of a Diels-Alder mechanophore. Using a
Min Zhang et al.
Journal of the American Chemical Society, 141(40), 15879-15883 (2019-09-07)
We have investigated the mechanical dissociation of an ammonium/crown ether rotaxane using experimental (sonication) and computational (CoGEF) methods and found that it breaks faster than its noninterlocked or uncoupled interlocked (i.e., pulled from both sides of the axle) counterparts. This
Guillaume De Bo et al.
Journal of the American Chemical Society, 139(25), 8455-8457 (2017-06-18)
We report on the synthesis of [2]rotaxanes driven by stabilization of the axle-forming transition state. A bifunctional macrocycle, with hydrogen bond donors at one end and acceptors at the other, is used to stabilize the charges that develop during the
Fubo Liang et al.
The Journal of biological chemistry, 278(43), 41734-41741 (2003-07-31)
Yersinia are causative agents in human diseases ranging from gastrointestinal syndromes to Bubonic Plague. There is increasing risk of misuse of infectious agents, such as Yersinia pestis, as weapons of terror as well as instruments of warfare for mass destruction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service