Skip to Content
Merck
All Photos(3)

Key Documents

274313

Sigma-Aldrich

Phloridzin dihydrate

99%

Synonym(s):

1-[2-(β-D-Glucopyranosyloxy)-4,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)-1-propanone, Phloretin 2′-β-D-glucopyranoside, Phloretin 2′-β-D-glucoside, Phlorizin dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€145.00
5 G
€691.00

€145.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€145.00
5 G
€691.00

About This Item

Empirical Formula (Hill Notation):
C21H24O10 · 2H2O
CAS Number:
Molecular Weight:
472.44
Beilstein:
66621
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.22

€145.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

optical activity

[α]20/D −52°, c = 3 in ethanol

mp

113-114 °C (lit.)

SMILES string

O.O.OC[C@H]1O[C@@H](Oc2cc(O)cc(O)c2C(=O)CCc3ccc(O)cc3)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H24O10.2H2O/c22-9-16-18(27)19(28)20(29)21(31-16)30-15-8-12(24)7-14(26)17(15)13(25)6-3-10-1-4-11(23)5-2-10;;/h1-2,4-5,7-8,16,18-24,26-29H,3,6,9H2;2*1H2/t16-,18-,19+,20-,21-;;/m1../s1

InChI key

XQWBNXSENPTIDY-YXMARJSJSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Used to induce experimental glycosuria.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rune E Kuhre et al.
Diabetologia, 62(6), 1011-1023 (2019-03-25)
Sodium-glucose cotransporter (SGLT) 2 inhibitors constitute a new class of glucose-lowering drugs, but they increase glucagon secretion, which may counteract their glucose-lowering effect. Previous studies using static incubation of isolated human islets or the glucagon-secreting cell line α-TC1 suggested that
Eric Dale Buras et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(8), 3537-3548 (2015-05-09)
Adipose tissue macrophages (ATMs) play an important role in the pathogenesis of obese type 2 diabetes. High-fat diet (HFD)-induced obesity has been shown to lead to ATM accumulation in rodents; however, the impact of hyperglycemia on ATM dynamics in HFD-fed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service