Skip to Content
Merck
All Photos(1)

Key Documents

273066

Sigma-Aldrich

4-Amino-2,6-diphenylphenol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€62.80

€62.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€62.80

About This Item

Linear Formula:
H2NC6H2(C6H5)2OH
CAS Number:
Molecular Weight:
261.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€62.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

148-150 °C (lit.)

functional group

phenyl

SMILES string

Nc1cc(c(O)c(c1)-c2ccccc2)-c3ccccc3

InChI

1S/C18H15NO/c19-15-11-16(13-7-3-1-4-8-13)18(20)17(12-15)14-9-5-2-6-10-14/h1-12,20H,19H2

InChI key

YCOUFOVMXBWYIX-UHFFFAOYSA-N

Related Categories

Application

4-Amino-2,6-diphenylphenol was used in the synthesis of 4-diazo-2,6-diphenylcyclohexa-2,5-dienone by undergoing diazotization in glacial acetic acid[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Equilibrium study of biphenyl-2-ol by infrared spectroscopy.
J. Chem. Soc. Perkin Trans. II, 14, 1663-1665 (1976)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service