Skip to Content
Merck
All Photos(1)

Key Documents

272469

Sigma-Aldrich

2-Cyanothioacetamide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€59.40
25 G
€165.00

€59.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€59.40
25 G
€165.00

About This Item

Linear Formula:
NCCH2CSNH2
CAS Number:
Molecular Weight:
100.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€59.40


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

118-120 °C (lit.)

functional group

amine
nitrile

SMILES string

NC(=S)CC#N

InChI

1S/C3H4N2S/c4-2-1-3(5)6/h1H2,(H2,5,6)

InChI key

BHPYMZQTCPRLNR-UHFFFAOYSA-N

Application

2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates[1] and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile][2]. It was also used as a building block for 2-pyridothiones[3][4].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liebigs Ann. Chem., 213-213 (1984)
Liebigs Ann. Chem., 210-210 (1986)
Farag M A Altalbawy
International journal of molecular sciences, 14(2), 2967-2979 (2013-02-01)
The title compounds were prepared by reaction of 1,1'-(5-methyl-1-phenyl-1H-pyrazole-3,4-diyl)diethanone (1) with different aromatic aldehydes 2a-c, namely Furfural (2a), 4-chlorobenzaldehyde (2b) and 4-methoxybenzaldhyde (2c) to yield the corresponding α,β-unsaturated ketones 3a-c. Compound 3 was reacted with malononitrile, 2-cyanoacetamide or 2-cyanothioacetamide yielded
A Krauze et al.
European journal of medicinal chemistry, 40(11), 1163-1167 (2005-06-02)
3,4-trans-4-Aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates 6-11 were prepared by a Michael reaction of N-acetonylpyridinium chloride with 3-aryl-2-cyanothioacrylamides or by a one-pot three-carbon condensation of N-acetonylpyridinium chloride, aromatic aldehyde and 2-cyanothioacetamide, and their cardiotonic properties were studied. 3,4-trans-5-cyano-2-hydroxy-2-methyl-4-(3-nitrophenyl)-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolate 8 was considered as a lead compound

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service