Skip to Content
Merck
All Photos(1)

Key Documents

271217

Supelco

Sephadex® G-150

for gel filtration, 40-120 μm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
23151817

grade

for gel filtration

particle size

40-120 μm

pore size

1-150 kDa fractionation range

Looking for similar products? Visit Product Comparison Guide

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhipeng Li et al.
International immunopharmacology, 14(2), 145-149 (2012-07-10)
The study is to investigate the effect of compound IMMLG5521, a coumarin derivative, on lung inflammation induced by Sephadex in rats. Sephadex led to massive granulomas, infiltration of neutrophils and eosinophils, the increase of TNF-α level in BALF and lung
Dae-Young Lee et al.
Journal of medicinal food, 15(4), 399-405 (2012-02-10)
Grains of sugary rice were extracted with 80% aqueous methanol, and the concentrated extracts were successively partitioned using ethyl acetate, n-butanol, and water. From the n-butanol fractions, four flavonoid glycosides were isolated through repeated silica gel, octadecyl silica gel, and
Lila Boulekbache-Makhlouf et al.
Phytochemical analysis : PCA, 24(2), 162-170 (2012-08-30)
Eucalyptus species are widely cultivated in Mediterranean regions. Moreover, plants of this family have been utilized for medicinal purposes. A number of studies have been devoted to the identification of eucalypt phenolics, all of them have focused on specific families
Olga N Solovjeva et al.
The protein journal, 31(2), 137-140 (2012-01-03)
Catalytic activity has been demonstrated for holotransketolase in the absence of free bivalent cations in the medium. The two active centers of the enzyme are equivalent in both the catalytic activity and the affinity for the substrates. In the presence
Ebru Akkemik et al.
Environmental toxicology and pharmacology, 34(3), 888-894 (2012-09-20)
The glutathione S-transferases (EC 2.5.1.18) were purified and characterized from turkey liver for the first time. The enzyme was purified 252.7-fold with a yield of 45%, with a specific activity of 164.31 U/mg from turkey liver. The purity of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service