Skip to Content
Merck
All Photos(1)

Key Documents

269522

Sigma-Aldrich

N-(4-Ethoxybenzylidene)-4-butylaniline

98%

Synonym(s):

4-Butyl-N-(4-ethoxybenzylidene)aniline, EBBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C6H4N=CHC6H4OC2H5
CAS Number:
Molecular Weight:
281.39
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid crystal

mp

38-44 °C (lit.)

SMILES string

CCCCc1ccc(cc1)\N=C\c2ccc(OCC)cc2

InChI

1S/C19H23NO/c1-3-5-6-16-7-11-18(12-8-16)20-15-17-9-13-19(14-10-17)21-4-2/h7-15H,3-6H2,1-2H3/b20-15+

InChI key

DBOAVDSSZWDGTH-HMMYKYKNSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Franklin et al.
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group, 8(2), 253-262 (1992-03-01)
A temperature probe based on the magnetic resonance properties of an encapsulated liquid crystal has been investigated. Large changes in magnetic resonance signals occur as the liquid crystal undergoes a phase transition from an anisotropic (nematic) state to the isotropic
Bibhuti B Das et al.
Solid state nuclear magnetic resonance, 33(3), 57-63 (2008-04-15)
An improved pulse sequence for SLF experiments based on the magic sandwich (MS) scheme for homo-nuclear dipolar decoupling is proposed. The sequence incorporates a double MS, both on I and S spins and has been named as EXE-MS2. The proposed
D Wróbel
Biophysical chemistry, 26(1), 91-99 (1987-04-01)
The resonance Raman (RR) spectra of chlorophyll (Chl) a, Chl b and pheophytin a dissolved in a liquid-crystalline MBBA + EBBA mixture were measured. The RR frequencies of chlorophylls in the liquid crystal (LC) are compared with those of solutions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service