Skip to Content
Merck
All Photos(1)

Key Documents

268380

Sigma-Aldrich

Stearic acid

≥98%

Synonym(s):

1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
Beilstein:
608585
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 173.7 °C)

Assay

≥98%

bp

361 °C (lit.)

mp

67-72 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ameya Prabhakar et al.
ACS omega, 4(13), 15567-15580 (2019-10-02)
The ability of ultrasound contrast agents to enhance the cell membrane permeability in response to an ultrasound pulse has unveiled avenues to facilitate the delivery of a higher intracellular payload at target sites. In light of the above, we report
Palle J Pedersen et al.
Journal of medicinal chemistry, 53(9), 3782-3792 (2010-04-22)
The design of retinoid phospholipid prodrugs is described based on molecular dynamics simulations and cytotoxicity studies of synthetic retinoid esters. The prodrugs are degradable by secretory phospholipase A(2) IIA and have potential in liposomal drug delivery targeting tumors. We have
Ulla Hoppu et al.
Journal of pediatric gastroenterology and nutrition, 41(3), 335-338 (2005-09-01)
Breast milk fatty acids may have immunomodulatory properties related to the development of atopic disease. The aim of this study was to assess the impact of the breast milk fatty acid composition on the development of atopic dermatitis (AD) in
Li Zeng et al.
PloS one, 3(6), e2329-e2329 (2008-06-05)
DNA damage triggers a network of signaling events that leads to cell cycle arrest or apoptosis. This DNA damage response acts as a mechanism to prevent cancer development. It has been reported that fatty acids (FAs) synthesis is increased in
Piret Arukuusk et al.
Biochimica et biophysica acta, 1828(5), 1365-1373 (2013-01-30)
Harnessing of a branched structure is a novel approach in the design of cell-penetrating peptides and it has provided highly efficient transfection reagents for intracellular delivery of nucleic acids. The new stearylated TP10 analogs, NickFects, condense plasmid DNA, splice correcting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service