Skip to Content
Merck
All Photos(1)

Key Documents

268127

Sigma-Aldrich

Diethylphosphonoacetic acid

95%

Synonym(s):

(Diethoxyphosphinyl)acetic acid, Diethyl carboxymethylphosphonate, NSC 272281

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€40.80
25 ML
€124.00

€40.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€40.80
25 ML
€124.00

About This Item

Linear Formula:
(C2H5O)2P(O)CH2CO2H
CAS Number:
Molecular Weight:
196.14
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€40.80


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.445 (lit.)

bp

150 °C/0.05 mmHg (lit.)

density

1.220 g/mL at 25 °C (lit.)

functional group

carboxylic acid
phosphonate

SMILES string

CCOP(=O)(CC(O)=O)OCC

InChI

1S/C6H13O5P/c1-3-10-12(9,11-4-2)5-6(7)8/h3-5H2,1-2H3,(H,7,8)

InChI key

DVQMPWOLBFKUMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acts as a nucleophile for nucleophilic addition reactions for synthesis of allene epoxides

Reactant for:
  • Stereoselectivity studies of the Staudinger reaction
  • Enantioselective formation of diols via epoxidation and hydration reactions
  • Horner-Wadsworth-Emmons reactions
  • Remote chelation controlled Ireland-Claisen rearrangement
  • Ugi-Dieckmann reactions for synthesis of tetramic acid derivatives
Used to make phosphonoester intermediates for intramolecular HEW reactions.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 113, 3850-3850 (1991)
Tetrahedron Asymmetry, 1, 137-137 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service