267538
Tungsten
foil, thickness 0.05 mm, ≥99.9% trace metals basis
Synonym(s):
W
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
≥99.9% trace metals basis
form
foil
resistivity
4.9 μΩ-cm, 20°C
thickness
0.05 mm
bp
5660 °C (lit.)
mp
3410 °C (lit.)
density
19.3 g/mL at 25 °C (lit.)
SMILES string
[W]
InChI
1S/W
InChI key
WFKWXMTUELFFGS-UHFFFAOYSA-N
Quantity
2.4 g = 50 × 50 mm; 9.6 g = 100 × 100 mm
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Denys Shevchenko et al.
Dalton transactions (Cambridge, England : 2003), 42(14), 5130-5139 (2013-02-14)
A rare example of a "monomeric" triple transition-metal substituted Keggin anion has been synthesized and characterized by various methods including X-ray crystallography, ESI and MALDI mass spectrometry, electrochemistry, EPR, and SQUID.
Karel Deprez et al.
Medical physics, 40(1), 012501-012501 (2013-01-10)
The construction of complex collimators with a high number of oblique pinholes is very labor intensive, expensive or is sometimes impossible with the current available techniques (drilling, milling or electric discharge machining). All these techniques are subtractive: one starts from
Zhigang Chen et al.
Advanced materials (Deerfield Beach, Fla.), 25(14), 2095-2100 (2013-02-22)
A new photothermal coupling agent for photothermal ablation (PTA) therapy of tumors is developed based on ultrathin PEGylated W18O49 nanowires. After being injected with the nanowire solution, the in vivo tumors exhibit a rapid temperature rise to 50.0 ± 0.5
Bing Sun et al.
Biosensors & bioelectronics, 44, 48-51 (2013-02-09)
A universal photoelectrochemical (PEC) sensing platform was fabricated based on the composition of protoporphyrin IX (PPIX), tungsten trioxide (WO3) and reduced graphene oxide (rGO) on indium tin oxide (ITO) electrode for detecting cysteine in aqueous solution. The rGO layer was
Weijie Zhao et al.
ACS nano, 7(1), 791-797 (2012-12-22)
Geometrical confinement effect in exfoliated sheets of layered materials leads to significant evolution of energy dispersion in mono- to few-layer thickness regime. Molybdenum disulfide (MoS(2)) was recently found to exhibit indirect-to-direct gap transition when the thickness is reduced to a
Articles
Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service