Skip to Content
Merck
All Photos(1)

Key Documents

26630

Sigma-Aldrich

2-Chloro-1,4-dimethylbenzene

≥99.0%

Synonym(s):

2-Chloro-p-xylene

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€204.00

€204.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€204.00

About This Item

Linear Formula:
(CH3)2C6H3Cl
CAS Number:
Molecular Weight:
140.61
Beilstein:
2040349
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€204.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99.0%

form

liquid

refractive index

n20/D 1.524 (lit.)
n20/D 1.524

bp

186 °C (lit.)

mp

2 °C (lit.)
2-3 °C

density

1.049 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Cc1ccc(C)c(Cl)c1

InChI

1S/C8H9Cl/c1-6-3-4-7(2)8(9)5-6/h3-5H,1-2H3

InChI key

KZNRNQGTVRTDPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-1,4-dimethylbenzene may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service