Skip to Content
Merck
All Photos(1)

Key Documents

265098

Sigma-Aldrich

Methyl 2,6-dihydroxy-4-methylbenzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H2(OH)2CO2CH3
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

96-98 °C (lit.)

solubility

chloroform: soluble 25 mg/mL, clear, colorless

functional group

ester

SMILES string

COC(=O)c1c(O)cc(C)cc1O

InChI

1S/C9H10O4/c1-5-3-6(10)8(7(11)4-5)9(12)13-2/h3-4,10-11H,1-2H3

InChI key

RIJMQNGJNNAAQK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T K Jayasekara et al.
Journal of mass spectrometry : JMS, 37(6), 577-580 (2002-07-12)
Securidaca longepedunculata Fers (Polygalaceae) is commonly used as a medicine in many parts of Africa and shows promise for protecting stored grain against insect pests. Analysis of a methanol extract of the root bark by gas chromatography linked to mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service