Skip to Content
Merck
All Photos(5)

Key Documents

264806

Sigma-Aldrich

(2,5-Dimethoxyphenyl)acetyl chloride

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€103.00
5 G
€405.00

€103.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€103.00
5 G
€405.00

About This Item

Linear Formula:
(CH3O)2C6H3CH2COCl
CAS Number:
Molecular Weight:
214.65
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€103.00


Estimated to ship on12 April 2025


Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.5385 (lit.)

bp

118 °C/0.8 mmHg (lit.)

density

1.227 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

COc1ccc(OC)c(CC(Cl)=O)c1

InChI

1S/C10H11ClO3/c1-13-8-3-4-9(14-2)7(5-8)6-10(11)12/h3-5H,6H2,1-2H3

InChI key

RAPIPBRHRYSAHQ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service