Skip to Content
Merck
All Photos(1)

Documents

263222

Sigma-Aldrich

Thulium

powder, ~40 mesh, 99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Tm
CAS Number:
Molecular Weight:
168.93
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: thulium

resistivity

90 μΩ-cm, 20°C

particle size

~40 mesh

bp

1950 °C (lit.)

mp

1545 °C (lit.)

density

9.332 g/mL at 25 °C (lit.)

SMILES string

[Tm]

InChI

1S/Tm

InChI key

FRNOGLGSGLTDKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burcu Tunc et al.
Lasers in surgery and medicine, 45(1), 48-56 (2013-01-17)
The thermal damage of the surrounding tissue can be an unwanted result of continuous-wave laser irradiations. In order to propose an effective alternative to conventional surgical techniques, photothermal damage must be taken under control by a detailed dose study. Real-time
Fabrizio Iacono et al.
BMC surgery, 12 Suppl 1, S21-S21 (2012-11-28)
Over the past years laser technology has played a predominant role in prostate surgery, for the treatment of benign prostate hypertrophy (BPH). Various laser devices have been introduced in clinical practice, showing good results in terms of complications and urodynamic
Zhonghua Yang et al.
Urology, 81(2), 396-400 (2013-02-05)
To compare the clinical outcomes between thulium laser transurethral enucleation of the prostate (ThuLEP) and plasmakinetic bipolar resection of the prostate (PKRP) for treating benign prostatic hyperplasia (BPH) in a prospective randomized trial with 18 months of follow-up. The study
Richard L Blackmon et al.
Journal of biomedical optics, 18(2), 28001-28001 (2013-02-05)
Fiber-optic attraction of urinary stones during laser lithotripsy may be exploited to manipulate stone fragments inside the urinary tract without mechanical grasping tools, saving the urologist time and space in the ureteroscope working channel. We compare thulium fiber laser (TFL)
Western Bolanos et al.
Optics letters, 37(19), 4032-4034 (2012-10-03)
Continuous wave laser operation at 1.87 μm of liquid-phase epitaxially (LPE) grown Tm(3+)-doped YLiF(4) (Tm:YLF) layers is demonstrated. The waveguide laser delivers 560 mW by pumping with a Ti:Sapphire laser at 780 nm leading to an efficiency of 76% with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service