Skip to Content
Merck
All Photos(1)

Key Documents

263036

Sigma-Aldrich

Dysprosium

powder, ~40 mesh, 99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Dy
CAS Number:
Molecular Weight:
162.50
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.9%

form

powder

resistivity

89 μΩ-cm, 20°C

particle size

~40 mesh

bp

2567 °C (lit.)

mp

1412 °C (lit.)

density

8.559 g/mL at 25 °C (lit.)

SMILES string

[Dy]

InChI

1S/Dy

InChI key

KBQHZAAAGSGFKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott A Sulway et al.
Chemical communications (Cambridge, England), 48(10), 1508-1510 (2011-09-20)
[Cp(2)Dy(thf)(μ-Cl)](2) (2) was synthesized from [Cp(2)Dy(μ-Cl)](n), which crystalizes as two polymorphs, with n = 2 (1a) or ∞ (1b). All three compounds show slow relaxation of magnetization, and in 2 the quantum tunnelling was found to be exchange-biased.
C Manjunatha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 140-148 (2012-04-07)
CdSiO(3):Dy(3+) (1-9mol%) nanophosphors were prepared for the first time using the solution combustion method. The process of monoclinic phase formation was investigated by PXRD, TG-DTA and FTIR. The results show that the phase formation temperature of combustion-derived monoclinic CdSiO(3) is
Magnetic anisotropy in a dysprosium/DOTA single-molecule magnet: beyond simple magneto-structural correlations.
Giuseppe Cucinotta et al.
Angewandte Chemie (International ed. in English), 51(7), 1606-1610 (2012-01-14)
Dimitris I Alexandropoulos et al.
Inorganic chemistry, 52(3), 1179-1181 (2013-01-18)
The first use of N-salicylidene-o-aminophenol in 3d/4f chemistry has led to Mn(III)(4)Dy(III)(5) and Mn(III)(4)Dy(III)(3) clusters with unprecedented metal topologies and stoichiometries; both compounds exhibit out-of-phase signals indicative of the slow magnetization relaxation of a single-molecule magnet.
Hai-Hong Wu et al.
Dalton transactions (Cambridge, England : 2003), 42(2), 342-346 (2012-11-15)
The heterometallic appended {MMn(III)(4)} (M = Dy(3+) and K(+)) cubanes were firstly trapped by two diamagnetic POM shells, which were robust enough to construct inorganic crystalline tubular materials. Magnetic study reveals the presence of a SMM-like slow magnetic relaxation feature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service