Skip to Content
Merck
All Photos(1)

Key Documents

262676

Sigma-Aldrich

Dichlorobis(triphenylphosphine)cobalt(II)

98%

Synonym(s):

Bis(triphenylphosphine)cobalt dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€159.00

€159.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€159.00

About This Item

Linear Formula:
[(C6H5)3P]2CoCl2
CAS Number:
Molecular Weight:
654.41
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€159.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

reaction suitability

core: cobalt
reagent type: catalyst

mp

236 °C (dec.) (lit.)

SMILES string

Cl[Co]Cl.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.2ClH.Co/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2

InChI key

OPRPFIIIFJLFCE-UHFFFAOYSA-L

Application

  • Catalyst in three-component coupling reactions
  • Catalyst for Epoxidation reactions
  • Catalyst in Hydrovinylation of Styrene
  • Catalyst in cross-coupling reactions
  • Catalyst for dimerization reactions
  • Catalyst in oxidation reactions

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service