Skip to Content
Merck
All Photos(1)

Documents

26253

Sigma-Aldrich

(3-Chloropropyl)trimethylsilane

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15ClSi
CAS Number:
Molecular Weight:
150.72
Beilstein:
1732625
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

refractive index

n20/D 1.431

bp

151 °C (lit.)

density

0.878 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)CCCCl

InChI

1S/C6H15ClSi/c1-8(2,3)6-4-5-7/h4-6H2,1-3H3

InChI key

QXDDDCNYAAJLBT-UHFFFAOYSA-N

Application

(3-Chloropropyl)trimethylsilane has been used in the preparation of:
  • 8-(3-trimethylsilylpropoxy)quinolone under phase transfer catalytic conditions
  • 5-trimethylsilyl pentanol
  • N-(tri-methylsilylalkyl)diamines

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silyl Modification of Biologically Active Compounds. 10. Lipid Type Organosilicon Derivatives of 8-Hydroxyquinoline and N-(2-Hydroxyethyl)-1, 2, 3, 4-tetrahydro (sila, iso) quinolines.
Segal I, et al.
Chemistry of Heterocyclic Compounds, 41(5), 613-624 (2005)
Organosilicon Compounds. Part V. Synthesis of N-(trimethylsilylalkyl) diamines and N-(trimethylsilylalkyl)-N'-(2-mercaptoethyl) diamines.
Thames SF and Edwards LH.
J. Chem. Soc. Sect. C, 2339-2342 (1968)
New deoxynojirimycin derivatives as potent inhibitors of intestinal a-glucohydrolases.
Lesur B, et al.
Bioorganic & Medicinal Chemistry Letters, 7(3), 355-360 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service