Skip to Content
Merck
All Photos(1)

Key Documents

261610

Sigma-Aldrich

Cyanogen bromide solution

5.0 M in acetonitrile

Synonym(s):

Bromine cyanide, Bromine monocyanide, Bromocyanide, Bromocyanogen, Carbononitridic bromide, Cyanic bromide, Cyanobromide, Cyanogen monobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCN
CAS Number:
Molecular Weight:
105.92
Beilstein:
1697296
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

5.0 M in acetonitrile

solubility

alcohol: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
water: freely soluble(lit.)

density

1.093 g/mL at 25 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

BrC#N

InChI

1S/CBrN/c2-1-3

InChI key

ATDGTVJJHBUTRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyanogen bromide solution induces template-guided condensation of oligonucleotides only in the presence of N-substituted morpholines. Mechanism of the phosphomonoester group activation by cyanogen bromide in N-substituted morpholine buffers has been investigated.

Application

Cyanogen bromide is a general reagent used to cleave carbon-heteroatom bonds. It is used in the synthesis of organocyanamides from corresponding tertiary amines, which is popularly known as von Braun reaction. CNBr solution is widely used in protein immobilization and cleavage.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bromalkylierte aromatische Amine. II. Mitteilung.
v Braun J, et al.
Ber. Deutsch. Chem. Ges., 51, 273-282 (1918)
Wen Qian et al.
Scientific reports, 10(1), 6470-6470 (2020-04-15)
The conjugation of polysaccharides with an effective carrier protein is critical for the development of effective bacterial polysaccharide vaccines. Therefore, the identification and optimization of carrier proteins to induce an effective immune response is necessary for developing a combined vaccine.
Cyanogen Bromide.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Immobilization of lipases on hydrophobic supports involves the open form of the enzyme.
Manoel EA, et al.
Enz. Microbiol. Technol., 71, 53-57 (2015)
Z A Shabarova et al.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life, 27(5-6), 555-566 (2001-09-07)
Cyanogen bromide has been found to induce the template-guided condensation of oligonucleotides only in the presence of N-substituted morpholines. Based on 31P, 1H and 13C NMR spectroscopy data, the mechanism of the phosphomonoester group activation by cyanogen bromide in N-substituted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service