Skip to Content
Merck
All Photos(1)

Key Documents

260819

Sigma-Aldrich

3-Methylbenzamide

99%

Synonym(s):

m-Toluamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CONH2
CAS Number:
Molecular Weight:
135.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

mp

94-96 °C (lit.)

SMILES string

Cc1cccc(c1)C(N)=O

InChI

1S/C8H9NO/c1-6-3-2-4-7(5-6)8(9)10/h2-5H,1H3,(H2,9,10)

InChI key

WGRPQCFFBRDZFV-UHFFFAOYSA-N

Application

3-Methylbenzamide may be used in chemical synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly A Finlayson et al.
Aquatic toxicology (Amsterdam, Netherlands), 207, 13-18 (2018-12-05)
Little is known about the effects of contaminants that accumulate in sea turtles. When in vivo exposure studies have ethical and logistical barriers, as is the case with sea turtles, in vitro tools can provide important information on the effects
Miriam Viviane Dias et al.
International immunopharmacology, 70, 324-337 (2019-03-11)
We investigated the effect in vitro and in vivo of doxycycline hyclate (Dx), a broad-spectrum antibiotic inhibitor of matrix metaloproteinases (MMPs), on adult Schistosoma mansoni worms and granulomatous liver inflammation in infected mice. Adult S. mansoni worms in culture treated
Daniel Molins-Delgado et al.
Journal of chromatography. A, 1544, 33-40 (2018-03-06)
Insect repellents (IRs) are a group of organic chemicals whose function is to prevent the ability of insects of landing in a surface. These compounds have been found in the environment and may pose a risk to non-target organisms. In
J E Song et al.
Experimental & applied acarology, 76(3), 355-363 (2018-10-22)
Acaricidal activities of the active component isolated from Melissa officinalis oil and its structural analogues against Tyrophagus putrescentiae were evaluated using fumigant and contact bioassays. The structure of 3,7-dimethyl-2,6-octadienal purified from M. officinalis oil was elucidated with EI-MS, 1H- and
Kerstin Büchel et al.
Ticks and tick-borne diseases, 6(4), 494-498 (2015-05-06)
Repellent efficacy of 10% EBAAP (3-[N-butyl-N-acetyl]-aminopropionic acid, ethyl ester) and 10% Icaridin ((2-(2-hydroxyethyl)-1-piperidinecarboxylic acid 1-methylpropyl ester)) were evaluated against 20% DEET (N,N-diethyl-3-methylbenzamide) in human subject trials against ticks. Responses of host-seeking nymphs of the European castor bean tick (Ixodes ricinus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service