Skip to Content
Merck
All Photos(1)

Key Documents

260541

Sigma-Aldrich

Glyoxal bis(diallyl acetal)

Synonym(s):

1,1,2,2-Tetraallyloxyethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(OCH2CH=CH2)2]2
CAS Number:
Molecular Weight:
254.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.4556 (lit.)

bp

155-160 °C/25 mmHg (lit.)

density

1.001 g/mL at 25 °C (lit.)

SMILES string

C=CCOC(OCC=C)C(OCC=C)OCC=C

InChI

1S/C14H22O4/c1-5-9-15-13(16-10-6-2)14(17-11-7-3)18-12-8-4/h5-8,13-14H,1-4,9-12H2

InChI key

BXAAQNFGSQKPDZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 38, 556-556 (1973)
THE INFLUENCE OF THIOL-ENE POLYMER EVOLUTION ON THE FORMATION AND PERFORMANCE OF HOLOGRAPHIC POLYMER DISPERSED LIQUID CRYSTALS.
White TJ, et al.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 47(2), 1013-1013 (2006)
Reza Zandi Shafagh et al.
ACS nano, 12(10), 9940-9946 (2018-09-14)
Electron beam lithography (EBL) is of major importance for ultraminiaturized biohybrid system fabrication, as it allows combining biomolecular patterning and mechanical structure definition on the nanoscale. Existing methods are limited by multistep biomolecule immobilization procedures, harsh processing conditions that are
Rheological properties of lightly crosslinked carboxy copolymers in aqueous solutions.
Nae HN and Reichert WW.
Rheologica Acta, 31(4), 351-360 (1992)
Network and swelling parameters of chemically crosslinked thermoreversible hydrogels.
Xue W, et al.
Polymer, 42(8), 3665-3669 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service