Skip to Content
Merck
All Photos(1)

Key Documents

260320

Sigma-Aldrich

1,2-Decanediol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€97.40

€97.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€97.40

About This Item

Linear Formula:
CH3(CH2)7CH(OH)CH2OH
CAS Number:
Molecular Weight:
174.28
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€97.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

255 °C (lit.)

mp

48-50 °C (lit.)

SMILES string

CCCCCCCCC(O)CO

InChI

1S/C10H22O2/c1-2-3-4-5-6-7-8-10(12)9-11/h10-12H,2-9H2,1H3

InChI key

YSRSBDQINUMTIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anisha Viswanathan et al.
European journal of pharmacology, 837, 105-116 (2018-09-05)
Glioblastoma remains the most common and aggressive type of malignant brain tumor among adults thus, considerable attention has been given to discovery of novel anti-tumor drugs for its treatment. This study reports the synthesis of a series of twelve novel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service