Skip to Content
Merck
All Photos(2)

Key Documents

259535

Sigma-Aldrich

Triethylenetetramine hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2(NHCH2CH2)2NH2 · xH2O
CAS Number:
Molecular Weight:
146.23 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

~5 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

98%

form

liquid

bp

272 °C (lit.)

mp

45-48 °C (lit.)

functional group

amine

storage temp.

−20°C

SMILES string

O.NCCNCCNCCN

InChI

1S/C6H18N4.H2O/c7-1-3-9-5-6-10-4-2-8;/h9-10H,1-8H2;1H2

InChI key

XJFQPBHOGJVOHU-UHFFFAOYSA-N

Related Categories

Application

Triethylenetetramine hydrate has been used as solvent to develop cellulose collector with immobilized triethylenetetraminepentaacetic acid groups for multielement preconcentration in flow system[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cellulose TETPA: a chelating collector designed for multielement preconcentration in flow systems.
Burba P, et al.
Fresenius Journal of Analytical Chemistry, 346(4), 414-419 (1993)
Thien Van Truong et al.
Colloids and surfaces. B, Biointerfaces, 193, 111101-111101 (2020-05-21)
We report the first observation of an efficient, native membrane conjugation mechanism via positively charged, linear oligo-amines. Clustering of membrane fragments relies on electrostatic interactions between the net negative charge of the membranes and the positively charged, water-soluble mediators. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service