Skip to Content
Merck
All Photos(1)

Key Documents

259365

Sigma-Aldrich

2,5-Difluorobenzoyl chloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€75.80

€75.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€75.80

About This Item

Linear Formula:
F2C6H3COCl
CAS Number:
Molecular Weight:
176.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€75.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.515 (lit.)

bp

92-93 °C/34 mmHg (lit.)

density

1.425 g/mL at 25 °C (lit.)

functional group

acyl chloride
fluoro

SMILES string

Fc1ccc(F)c(c1)C(Cl)=O

InChI

1S/C7H3ClF2O/c8-7(11)5-3-4(9)1-2-6(5)10/h1-3H

InChI key

RLRUKKDFNWXXRT-UHFFFAOYSA-N

Application

2,5-Difluorobenzoyl chloride has been used in the synthesis of:
  • ethyl 4-(2′,5′-difluorobenzoyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate (JJ78:1)[1]
  • thiohydrazide[2]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, determination of absolute configuration, and biological evaluation of spiro-fused thiadiazoline inhibitors of kinesin spindle protein (KSP).
Angeles AR, et al.
Tetrahedron Letters, 51(48), 6236-6239 (2010)
Oliver D Staples et al.
Cell cycle (Georgetown, Tex.), 7(21), 3417-3427 (2008-10-31)
A robust p53 cell-based assay that exploits p53's function as a transcription factor was used to screen a small molecule library and identify bioactive small molecules with potential antitumor activity. Unexpectedly, the majority of the highest ranking hit compounds from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service