Skip to Content
Merck
All Photos(1)

Key Documents

256579

Sigma-Aldrich

3,3,3-Triphenylpropionic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€82.90

€82.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€82.90

About This Item

Linear Formula:
(C6H5)3CCH2CO2H
CAS Number:
Molecular Weight:
302.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€82.90


Check Cart for Availability

Request a Bulk Order

vapor density

10.1 (vs air)

Assay

97%

mp

180-182 °C (lit.)

functional group

carboxylic acid
phenyl

SMILES string

OC(=O)CC(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H18O2/c22-20(23)16-21(17-10-4-1-5-11-17,18-12-6-2-7-13-18)19-14-8-3-9-15-19/h1-15H,16H2,(H,22,23)

InChI key

XMSJLUKCGWQAHO-UHFFFAOYSA-N

General description

Reaction of 3,3,3-triphenylpropionic acid with lead tetraacetate in benzene, acetonitrile or chlorobenzene solution has been investigated[1].

Application

3,3,3-Triphenylpropionic acid has been used in the preparation of trityl-deprotected aminoalkoxysilane[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concurrent Carbon-to-Oxygen Rearrangement, Cyclization, and Decarboxylation in the Reaction of 3, 3, 3-Triarylpropionic Acids with Lead Tetraacetate.
Starnes WH.
Journal of the American Chemical Society, 86(24), 5603-5611 (1964)
Spacing and site isolation of amine groups in 3-aminopropyl-grafted silica materials: The role of protecting groups.
Hicks JC, et al.
Chemistry of Materials, 18(21), 5022-5032 (2006)
Quentin Duez et al.
Polymers, 11(4) (2019-04-19)
Several families of polymers possessing various end-groups are characterized by ion mobility mass spectrometry (IMMS). A significant contribution of the end-groups to the ion collision cross section (CCS) is observed, although their role is neglected in current fitting models described

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service