Skip to Content
Merck
All Photos(2)

Key Documents

256323

Sigma-Aldrich

(S)-(−)-1-Phenyl-1-propanol

99%

Synonym(s):

(S)-(−)-α-Ethylbenzyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€451.00

€451.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€451.00

About This Item

Linear Formula:
C2H5CH(C6H5)OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
2041556
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

€451.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

optical activity

[α]20/D −47°, c = 2.25 in hexane

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.520 (lit.)

bp

94-95 °C/10 mmHg (lit.)

density

0.992 g/mL at 25 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

CC[C@H](O)c1ccccc1

InChI

1S/C9H12O/c1-2-9(10)8-6-4-3-5-7-8/h3-7,9-10H,2H2,1H3/t9-/m0/s1

InChI key

DYUQAZSOFZSPHD-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral building block.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hisashi Matsuda et al.
Bioorganic & medicinal chemistry letters, 15(7), 1949-1953 (2005-03-23)
1'S-1'-Acetoxychavicol acetate from the rhizomes of Alpinia galanga inhibited nitric oxide (NO) production in lipopolysaccharide-activated mouse peritoneal macrophages with an IC(50) value of 2.3 microM. To clarify the structure-activity relationship of 1'S-1'-acetoxychavicol acetate, various natural and synthetic phenylpropanoids and synthetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service