Skip to Content
Merck
All Photos(3)

Documents

255637

Sigma-Aldrich

Iron

flakes, ≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.99% trace metals basis

form

flakes

resistivity

9.71 μΩ-cm

diam. × thickness

<25 mm × 4-6 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Daily iron during pregnancy improves birth weight.
Jacqui Wise
BMJ (Clinical research ed.), 346, f3997-f3997 (2013-06-26)
Tomasz Pytrus et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 34(203), 263-268 (2013-07-31)
The aim of the study was to analyze the occurrence of anemia in children with newly diagnosed Crohn's disease and 3 months after the initiation of treatment depending on location and activity of the disease. 36 children aged from 2
Elzbieta Krzesiek et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 34(203), 289-292 (2013-07-31)
Anemia is a frequent symptom of diseases of alimentary tract, also in children. Among others, inflammatory bowel disease, celiac disease and Helicobacter pylori are most often complicated by anemia. Not infrequently these disorders are accompanied by more than one type
Stanley Zlotkin et al.
JAMA, 310(9), 938-947 (2013-09-05)
In sub-Saharan Africa, malaria is a leading cause of childhood morbidity and iron deficiency is among the most prevalent nutritional deficiencies. In 2006, the World Health Organization and the United Nations Children's Fund released a joint statement that recommended limiting
Shaun D Wong et al.
Nature, 499(7458), 320-323 (2013-07-23)
Mononuclear non-haem iron (NHFe) enzymes catalyse a broad range of oxidative reactions, including halogenation, hydroxylation, ring closure, desaturation and aromatic ring cleavage reactions. They are involved in a number of biological processes, including phenylalanine metabolism, the production of neurotransmitters, the

Articles

An article concerning self-propagating reactions induced by mechanical alloying, presented by Sigma-Aldrich.com.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service