Skip to Content
Merck
All Photos(1)

Key Documents

255335

Sigma-Aldrich

2-Thiophenesulfonyl chloride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3ClO2S2
CAS Number:
Molecular Weight:
182.65
Beilstein:
123536
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

bp

130-132 °C/14 mmHg (lit.)

mp

30-32 °C (lit.)

SMILES string

ClS(=O)(=O)c1cccs1

InChI

1S/C4H3ClO2S2/c5-9(6,7)4-2-1-3-8-4/h1-3H

InChI key

VNNLHYZDXIBHKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics of reactions of 2-thiophenesulfonyl chloride with substituted anilines in methanol has been reported[1]. Kinetics of solvolysis of 2-thiophenesulfonyl chloride in binary solvent mixtures has been studied[2].

Application

2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene[3].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-(trans-. beta.-Styrylsulfonyl)-and 2-((trans-2-phenylcyclopropyl) sulfonyl) thiophene.
Goralski CT.
The Journal of Organic Chemistry, 37(14), 2354-2355 (1972)
Malcolm J D'Souza et al.
International journal of molecular sciences, 9(5), 914-925 (2009-03-28)
Specific rates of solvolysis in hydroxylic solvents available for the solvolysis of 2-thiophenesulfonyl chloride and phenylmethanesulfonyl chloride are supplemented by determining the values in fluoroalcohol-containing solvents. The data sets are then correlated using the extended Grunwald-Winstein equation. For both substrates
Reaction kinetics of 3-thiophenesulfonyl chloride with anilines in methanol.
Arcoria A, et al.
The Journal of Organic Chemistry, 39(12), 1689-1691 (1974)
Radoslaw Laufer et al.
Bioorganic & medicinal chemistry, 22(17), 4968-4997 (2014-07-22)
TTK kinase was identified by in-house siRNA screen and pursued as a tractable, novel target for cancer treatment. A screening campaign and systematic optimization, supported by computer modeling led to an indazole core with key sulfamoylphenyl and acetamido moieties at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service