All Photos(1)
About This Item
Linear Formula:
O2NC6H4COC6H5
CAS Number:
Molecular Weight:
227.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
mp
92-94 °C (lit.)
SMILES string
[O-][N+](=O)c1cccc(c1)C(=O)c2ccccc2
InChI
1S/C13H9NO3/c15-13(10-5-2-1-3-6-10)11-7-4-8-12(9-11)14(16)17/h1-9H
InChI key
MFYLRNKOXORIPK-UHFFFAOYSA-N
Application
3-Nitrobenzophenone was used as starting reagent in the synthesis of ketoprofen, non-steroidal anti inflammatory drug. It was also used as plastic additive and its role in photolytic degradation of polyethylene and polypropylene in solution has been investigated.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Acetophenone and benzophenone derivatives as catalysts in photodegradation of PE and PP films.
Manangan T, et al.
Advanced Materials Research, 93, 284-287 (2010)
A comparative study on the photo-induced arylation of β-dicarbonyl compounds by arylazosulfides and its use in the synthesis of methyl labeled 2-arylpropionic acids.
Tona M, et al.
Tetrahedron, 50(27), 8117-8126 (1994)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service