Skip to Content
Merck
All Photos(1)

Key Documents

253421

Sigma-Aldrich

3-(2-Iodoacetamido)-PROXYL

free radical

Synonym(s):

3-(2-Iodoacetamido)-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18IN2O2
CAS Number:
Molecular Weight:
325.17
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

162-164 °C (lit.)

functional group

amide
iodo

storage temp.

2-8°C

SMILES string

CC1(C)CC(NC(=O)CI)C(C)(C)N1[O]

InChI

1S/C10H18IN2O2/c1-9(2)5-7(12-8(14)6-11)10(3,4)13(9)15/h7H,5-6H2,1-4H3,(H,12,14)

InChI key

OQIGMSGDHDTSFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(2-Iodoacetamido)-PROXYL is a free radical used as a spin probe in spin labeling techniques. This technique is helpful in investigating the dynamic aspects of molecular interactions of substances, proteins, lipids, and cell membranes.

Application

Sulfhydryl group specific spin label. Applications include study of ozone-treated human and bovine erythrocyte membrane proteins and the orientation of myosin subfragment on muscle fibers. DNA fragments containing phosphorothioate diesters can be labeled by spin labels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Li et al.
Scientific reports, 10(1), 393-393 (2020-01-17)
Fowlpox virus resolvase (Fpr) is an endonuclease that cleaves a broad range of branched DNA structures, including the Holliday junction (HJ), with little sequence-specificity. To better understand the mechanisms underlying its relaxed substrate specificity, we determined the crystal structures of
Christoph Gmeiner et al.
Physical chemistry chemical physics : PCCP, 19(41), 28360-28380 (2017-10-17)
A combined method, employing NMR and EPR spectroscopies, has demonstrated its strength in solving structures of protein/RNA and other types of biomolecular complexes. This method works particularly well when the large biomolecular complex consists of a limited number of rigid
Site directed nitroxide spin labeling of oligonucleotides for NMR and EPR studies
Shepherd NE, et al.
Tetrahedron, 71(5), 813-819 (2015)
Spin labeled calmodulin: a new probe for studying Ca2+ and macromolecular interactions.
P B Hewgley et al.
Annals of the New York Academy of Sciences, 356, 20-32 (1980-01-01)
Isabelle Lebars et al.
Nucleic acids research, 42(15), e117-e117 (2014-07-02)
Site-directed spin labeling is emerging as an essential tool to investigate the structural and dynamical features of RNA. We propose here an enzymatic method, which allows the insertion of a paramagnetic center at a specific position in an RNA molecule.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service