Skip to Content
Merck
All Photos(1)

Key Documents

253154

Sigma-Aldrich

2-Bromophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€64.70

€64.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€64.70

About This Item

Linear Formula:
BrC6H4NCS
CAS Number:
Molecular Weight:
214.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€64.70


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.6843 (lit.)

bp

257 °C/770 mmHg (lit.)

density

1.591 g/mL at 25 °C (lit.)

functional group

bromo
isothiocyanate

SMILES string

Brc1ccccc1N=C=S

InChI

1S/C7H4BrNS/c8-6-3-1-2-4-7(6)9-5-10/h1-4H

InChI key

PAFORXDSYWMYGP-UHFFFAOYSA-N

Application

2-Bromophenyl isothiocyanate has been used in the synthesis of 4-monosubstituted and 4,4-disubstituted 1,4-dihydro-3,1-benzoxazine-2-thiones[1].

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-pot synthesis of 1, 4-dihydro-3, 1-benzoxazine-2-thiones by the reaction of 2-lithiophenyl isothiocyanates with aldehydes or ketones.
Kobayashi K, et al.
Tetrahedron, 66(48), 9336-9339 (2010)
Fazila Rizvi et al.
Scientific reports, 9(1), 6738-6738 (2019-05-03)
A library of thiosemicarbazide derivatives of isoniazid 3-27, was synthesized and evaluated for their anti-inflammatory and urease inhibition activities, by using in vitro bioassays. Among these compounds 9, 10, 12, 21, and 26 were identified as new derivatives. Prolonged use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service