Skip to Content
Merck
All Photos(1)

Key Documents

25205

Sigma-Aldrich

Chloromethyl phenyl sulfoxide

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SOCH2Cl
CAS Number:
Molecular Weight:
174.65
Beilstein:
1617664
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0%

form

solid

bp

109-111 °C/0.01 mmHg

mp

33-35 °C

storage temp.

−20°C

SMILES string

ClCS(=O)c1ccccc1

InChI

1S/C7H7ClOS/c8-6-10(9)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

WCUSVVDFUIMPKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloromethyl phenyl sulfoxide has synthetic potential and exhibits unusual stereochemical specificity. Stereospecific hydroxyalkylation of chloromethyl phenyl sulfoxide has been investigated.

Application

Chloromethyl phenyl sulfoxide has been used in the preparation of:
  • 1-chloroalkyl sulfoxides
  • starting material for the synthesis alkyl sulfoxides and thiol ester acyl anion equivalent

Other Notes

Starting material for the synthesis alkyl sulfoxides; Thiol ester acyl anion equivalent

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.M. Moore et al.
The Journal of Organic Chemistry, 43, 2713-2713 (1978)
Tsuyoshi Satoh
Chemical record (New York, N.Y.), 3(6), 329-341 (2004-03-03)
Aryl 1-chlorovinyl sulfoxides were easily synthesized from ketones and aldehydes with aryl chloromethyl sulfoxide in three-steps with high overall yields. Low-temperature treatment of the aryl 1-chlorovinyl sulfoxides with alkyllithium or a Grignard reagent gave alkylidene carbenoids via a sulfoxide-metal exchange
M. Hojo et al.
Synthesis, 789-789 (1977)
Stereospecific hydroxyalkylation of chloromethyl phenyl sulfoxide.
Durst T.
Journal of the American Chemical Society, 91(4), 1034-1035 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service