Skip to Content
Merck
All Photos(1)

Documents

251984

Sigma-Aldrich

1-Iodooctadecane

95%

Synonym(s):

Octadecyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17I
CAS Number:
Molecular Weight:
380.39
Beilstein:
1755880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

bp

194-197 °C/2 mmHg (lit.)

mp

33-35 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCI

InChI

1S/C18H37I/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1H3

InChI key

ZNJOCVLVYVOUGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective adsorption of copper phthalocyanine atop functionalized organic monolayers.
Lei S, et al.
The Journal of Physical Chemistry B, 108(1), 224-227 (2004)
Controlled assembly of copper phthalocyanine with 1-iodooctadecane.
Lei S, et al.
Chinese Science Bulletin, 48(15), 1519-1524 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service