Skip to Content
Merck
All Photos(1)

Key Documents

251313

Sigma-Aldrich

Pimeloyl chloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€104.00

€104.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€104.00

About This Item

Linear Formula:
ClCO(CH2)5COCl
CAS Number:
Molecular Weight:
197.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€104.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.469 (lit.)

bp

113 °C/5 mmHg (lit.)

density

1.205 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

ClC(=O)CCCCCC(Cl)=O

InChI

1S/C7H10Cl2O2/c8-6(10)4-2-1-3-5-7(9)11/h1-5H2

InChI key

LVIMBOHJGMDKEJ-UHFFFAOYSA-N

Application

Pimeloyl chloride has been used in preparation of:
  • bis-acylureas, by the coupling reaction with ureas[1]
  • triacetyl-15-pimelate-nivalenol, required for production of antibody against nivalenol tetraacetate[2]
  • macrocyclic tetralactones, via condensation with stannoxane[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong-Uk Kim et al.
Journal of colloid and interface science, 359(2), 428-435 (2011-05-04)
We present the synthesis and morphology study of thirteen bis-acylurea molecules with various functional end groups. The bis-acylureas have two acylurea groups, -NH-CO-NH-CO-, divided by a pentamethylene spacer, -(CH(2))(5)-, and two symmetric functional end groups, such as, aliphatic, benzyl, mono-
Tin derivatives for synthesis: the preparation of macrocyclic lactones.
Shanzer A and Mayer-Shochet N.
Journal of the Chemical Society. Chemical Communications, 4, 176-177 (1980)
C R Wang et al.
Applied and environmental microbiology, 57(4), 1026-1030 (1991-04-01)
Antibody against nivalenol tetraacetate (tetra-Ac-NIV) was prepared by immunization of rabbits with triacetyl-15-pimelate-NIV conjugated to bovine serum albumin. By using tritiated tetra-Ac-NIV as the test ligand, antibody titers were demonstrated as early as 4 weeks after immunization. Useful antibody for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service