Skip to Content
Merck
All Photos(1)

Key Documents

251046

Sigma-Aldrich

α,α-Dichlorotoluene

95%

Synonym(s):

(Dichloromethyl)benzene, Benzal chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHCl2
CAS Number:
Molecular Weight:
161.03
Beilstein:
1099407
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
95%

vapor pressure

0.3 mmHg ( 20 °C)

Assay

95%

form

liquid

refractive index

n20/D 1.550 (lit.)

bp

82 °C/10 mmHg (lit.)

mp

−17-−15 °C (lit.)

solubility

alcohol: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
water: insoluble(lit.)

density

1.254 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)c1ccccc1

InChI

1S/C7H6Cl2/c8-7(9)6-4-2-1-3-5-6/h1-5,7H

InChI key

CAHQGWAXKLQREW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vibrational spectra of α,α-dichlorotoluene has been studied in the range 3600-100 cm-1.

Application

α,α-Dichlorotoluene has been useds as initiator during the atom transfer radical polymerization (ATRP) of styrene and methyl methacrylate.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P B Nagabalasubramanian et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 73(2), 277-280 (2009-04-07)
The vibrational spectra of alpha,alpha dichlorotoluene have been recorded using PerkinElmer 521 spectrometer in the range 3600-100 cm(-1). A detailed vibrational spectral analysis has been carried out and assignments of the observed fundamental bands have been proposed on the basis
Atom transfer radical polymerization (ATRP) of styrene and methyl methacrylate with a, a-dichlorotoluene as initiator; a kinetic study.
Neumann A, et al.
Macromolecular Chemistry and Physics, 201(9), 980-984 (2000)
Benzal chloride.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 29, 65-72 (1982-05-01)
Occupational exposure to benzyl chloride and benzyl chloride due to contaminated butyl benzyl phthalate.
L Rittfeldt et al.
Scandinavian journal of work, environment & health, 9(4), 367-368 (1983-08-01)
Carlos Eduardo Verdiani de Carvalho et al.
Acta cirurgica brasileira, 21(1), 43-46 (2006-02-24)
To investigate the effects of intrinsic denervation of the jejunum after the extensive intestinal resection in rats. Thirty male Wistar rats were distributed into three groups, depending on the experimental procedure: Group C (control), Group R (resection) and Group D

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service