Skip to Content
Merck
All Photos(1)

Key Documents

251038

Sigma-Aldrich

2-Benzyl-2-thiopseudourea hydrochloride

98%

Synonym(s):

S-Benzylisothiourea hydrochloride, S-Benzylthiuronium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€108.00

€108.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€108.00

About This Item

Linear Formula:
C6H5CH2SC(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
202.70
Beilstein:
3656719
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€108.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

177-179 °C (lit.)

solubility

methanol: soluble 1 g/10 mL, clear, colorless to very faintly yellow

functional group

amine
phenyl
thioether

SMILES string

Cl.NC(=N)SCc1ccccc1

InChI

1S/C8H10N2S.ClH/c9-8(10)11-6-7-4-2-1-3-5-7;/h1-5H,6H2,(H3,9,10);1H

InChI key

WJAASTDRAAMYNK-UHFFFAOYSA-N

Related Categories

General description

S-benzylisothiourea analogs (2-benzyl-2-thiopseudourea hydrochloride), as small-molecule inhibitors of indoleamine-2,3-dioxygenase, has been studied[1].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Matsuno et al.
Bioorganic & medicinal chemistry letters, 20(17), 5126-5129 (2010-08-07)
S-benzylisothiourea 3a was discovered by its ability to inhibit indoleamine-2,3-dioxygenase (IDO) in our screening program. Subsequent optimization of the initial hit 3a lead to the identification of sub-muM inhibitors 3r and 10h, both of which suppressed kynurenine production in A431

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service