Skip to Content
Merck
All Photos(1)

Key Documents

250767

Sigma-Aldrich

Trichloromethyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl3NCO
CAS Number:
Molecular Weight:
160.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.477 (lit.)

bp

119-123 °C (lit.)

density

1.564 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(Cl)N=C=O

InChI

1S/C2Cl3NO/c3-2(4,5)6-1-7

InChI key

OXBFBRZWBIXFGO-UHFFFAOYSA-N

Application

Reactant involved in synthesis of:
  • Trisubstituted alkanoates or lactone derivatives
  • Urea and thiourea derivatives of 15-membered azalides for use as antimalarial chemotherapeutics
Trichloromethyl isocyanate has been used in derivatization of carboxy end functional poly(methylmethacrylate) or amino end function polystyrene (polymers)[1].

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in RAFT polymerization: the synthesis of polymers with defined end-groups.
Moad G, et al.
Polymer, 46(19), 8458-8468 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service