Skip to Content
Merck
All Photos(1)

Key Documents

249777

Sigma-Aldrich

(Bromomethyl)trimethylsilane

97%

Synonym(s):

(Trimethylsilyl)methyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2Br
CAS Number:
Molecular Weight:
167.12
Beilstein:
1731484
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

115.5 °C/742 mmHg (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)CBr

InChI

1S/C4H11BrSi/c1-6(2,3)4-5/h4H2,1-3H3

InChI key

ACAUYCZBWABOLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service