Skip to Content
Merck
All Photos(1)

Key Documents

249262

Sigma-Aldrich

(−)-Corey lactone, 4-phenylbenzoate alcohol

99%

Synonym(s):

(3aα,4α,5β,6aα)-(−)-Hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl 1,1′-biphenyl-4-carboxylate, (3aR,4S,5R,6aS)-Hexahydro-4-hydroxymethyl-5-(4-phenylbenzoyloxy)cyclopenta[b]furan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O5
CAS Number:
Molecular Weight:
352.38
Beilstein:
1294692
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]20/D −87°, c = 1 in chloroform

mp

131-132 °C (lit.)

functional group

ester
hydroxyl
phenyl

storage temp.

2-8°C

SMILES string

OC[C@H]1[C@@H](C[C@@H]2OC(=O)C[C@H]12)OC(=O)c3ccc(cc3)-c4ccccc4

InChI

1S/C21H20O5/c22-12-17-16-10-20(23)25-18(16)11-19(17)26-21(24)15-8-6-14(7-9-15)13-4-2-1-3-5-13/h1-9,16-19,22H,10-12H2/t16-,17-,18+,19-/m1/s1

InChI key

SZJVIFMPKWMGSX-AKHDSKFASA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Corey lactone, 4-phenyl benzoate alcohol, also known as 2H-cyclopenta[b]furan, [1,1′-biphenyl]-4-carboxylic acid derivative, is commonly used as a chiral substrate and is a versatile building block for prostaglandins. [1]

Application

(−)-Corey lactone, 4-phenyl benzoate alcohol is used as:
  • A substrate in the synthesis of tricyclic cyclopent[b]benzofuran. [2]
  • A Key intermediate in the synthesis of prostaglandins.[3][4]
  • Building block for a potent and selective antiglaucoma agent analog of PGF[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pot and time economies in the total synthesis of Corey lactone
Nariyoshi U,et al.
Chemical Science, 11, 1205-1209 (2020)
A practical synthesis of chiral tricyclic cyclopenta [b] benzofuran, a key intermediate of Beraprost
Xiaoyu L,et al.
Organic & Biomolecular Chemistry, 14, 7715-7721 (2016)
Aldrichimica Acta, 15, 65-65 (1982)
B Resul et al.
Journal of medicinal chemistry, 36(2), 243-248 (1993-01-22)
A series of phenyl-substituted analogues of prostaglandin F2 alpha (PGF2 alpha) were prepared and evaluated for ocular hypotensive effect and side effects in different animal models. In addition, the activity of the analogues on FP receptors was studied in vitro.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service