Skip to Content
Merck
All Photos(1)

Key Documents

247197

Sigma-Aldrich

Methyl vinyl sulfone

≥97%

Synonym(s):

(Methylsulfonyl)ethene, 1-Methylsulfonylethene, 1-Methylsulfonylethylene, Methanesulfonylethene, Methyl vinyl sulphone, Vinyl methyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHSO2CH3
CAS Number:
Molecular Weight:
106.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

contains

0.02% p-tert-butylphenol as stabilizer

refractive index

n20/D 1.463 (lit.)

bp

115-117 °C/19 mmHg (lit.)

density

1.212 g/mL at 25 °C (lit.)

functional group

sulfone

storage temp.

2-8°C

SMILES string

CS(=O)(=O)C=C

InChI

1S/C3H6O2S/c1-3-6(2,4)5/h3H,1H2,2H3

InChI key

WUIJTQZXUURFQU-UHFFFAOYSA-N

Gene Information

Application

Methyl vinyl sulfone has been used:
  • as derivatization reagent in an improved method for detection of pseudouridine in nucleoside mixtures by capillary HPLC-mass spectrometry[1]
  • in preparation of methylsulfonylethyl cellulose via Michael addition to cotton fabric[2]
  • in Heck vinylation of aryl halides[3]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gert Emmerechts et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 825(2), 233-238 (2005-07-21)
A method is presented for improved detection of pseudouridine in nucleoside mixtures based on the specific derivatization with methyl vinyl sulfone followed by analysis by capillary HPLC-mass spectrometry. Reaction conditions were optimized in order to obtain the best yield and
G B Gordon et al.
Carcinogenesis, 12(12), 2393-2396 (1991-12-01)
The induction of quinone reductase [QR; NAD(P)H:(quinone acceptor) oxidoreductase; EC 1.6.99.2] in cultured cells and animal tissues of rodents has provided useful information on mechanisms of protection against carcinogens. We have developed a simple and efficient microtiter plate assay for
Synthesis, 3495-3495 (2006)
K L Dearfield et al.
Mutagenesis, 6(6), 519-525 (1991-11-01)
Over the past several years, we have been evaluating the mutagenicity and clastogenicity of compounds capable of Michael-type reactions. These compounds, including acrylamide, several acrylate and methacrylate esters, vinyl sulfones, and phorone, have been evaluated using TK+/- -3.7.2C mouse lymphoma
M D Shelby et al.
Mutation research, 250(1-2), 431-437 (1991-09-01)
Methyl vinyl sulfone and divinyl sulfone were tested for the induction of dominant lethal mutations and micronucleated bone-marrow erythrocytes in male mice. These chemicals were chosen for study because of their similarities in structure and chemical reactivity to acrylamide which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service