Skip to Content
Merck
All Photos(1)

Key Documents

245623

Sigma-Aldrich

1-Chlorohexadecane

95%

Synonym(s):

Cetyl chloride, Hexadecyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€80.10

€80.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€80.10

About This Item

Linear Formula:
CH3(CH2)15Cl
CAS Number:
Molecular Weight:
260.89
Beilstein:
1748498
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€80.10


Check Cart for Availability

Request a Bulk Order

vapor pressure

129 mmHg ( 21 °C)

Assay

95%

form

liquid

refractive index

n20/D 1.449 (lit.)

bp

149 °C/1 mmHg (lit.)

mp

8-14 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCl

InChI

1S/C16H33Cl/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-16H2,1H3

InChI key

CLWAXFZCVYJLLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The electron impact mass spectra of 1-chlorohexadecane was studied by sampling with supersonic molecular beams[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Dagan et al.
Journal of the American Society for Mass Spectrometry, 6(2), 120-131 (1995-02-01)
The electron impact mass spectrometry of straight chain alkanes C8H18-C40H82, squalane, methylstearate, 1-chlorohexadecane, 1-bromohexadecane, and dioctylphthalate was studied by sampling them with supersonic molecular beams. A fly-through Brink-type electron impact ion source was used, utilizing a vacuum background ion filtration
K Koike et al.
Applied and environmental microbiology, 65(12), 5636-5638 (1999-12-03)
A mutant Rhodococcus strain lacking the ability to utilize 1-chlorohexadecane was found to cis-desaturate aliphatic compounds, such as 1-chlorohexadecane, n-hexadecane, and heptadecanonitrile, yielding corresponding products with a double bond mainly at the ninth carbon from the terminal methyl groups. A
G L Murphy et al.
Applied and environmental microbiology, 53(1), 10-13 (1987-01-01)
The composition of phospholipids from Mycobacterium convolutum R22 was determined after growth at two temperatures (20 and 30 degrees C) with 1-chlorohexadecane as the substrate. Comparisons were made with the phospholipids of cells grown on n-hexadecane. Phosphatidylinositolmannosides and phosphatidylethanolamine (PE)
Nicolas Goujon et al.
Physical chemistry chemical physics : PCCP, 17(35), 23059-23068 (2015-08-15)
The development of new polymerizable lyotropic liquid crystals (LLCs) utilizing charged amphiphilic molecules such as those based on long chain imidazolium compounds, is a relatively new design direction for producing robust membranes with controllable nano-structures. Here we have developed a
G L Murphy et al.
Journal of bacteriology, 156(3), 1158-1164 (1983-12-01)
The cellular fatty acid composition of Mycobacterium vaccae JOB5 and Mycobacterium convolutum R22 was examined after growth on n-alkanes and compared with the fatty acids of the organisms after growth on 1-chlorohexadecane and 1-chlorooctadecane. Growth on n-alkanes resulted in normal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service