Skip to Content
Merck
All Photos(1)

Key Documents

244872

Sigma-Aldrich

Sodium-potassium alloy

Potassium 78 wt. %, Sodium 22 wt. %

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€324.00

€324.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€324.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

€324.00


Estimated to ship on24 May 2025


Request a Bulk Order

form

liquid

composition

Potassium, 78 wt. %
Sodium, 22 wt. %

reaction suitability

reagent type: reductant

SMILES string

[Na].[K]

InChI

1S/K.Na

InChI key

BITYAPCSNKJESK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service