Skip to Content
Merck
All Photos(1)

Documents

244198

Sigma-Aldrich

1-(2,4,6-Triisopropylbenzenesulfonyl)-3-nitro-1H-1,2,4-triazole

Synonym(s):

TPSNI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24N4O4S
CAS Number:
Molecular Weight:
380.46
Beilstein:
585765
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

mp

128-130 °C (lit.)

SMILES string

CC(C)c1cc(C(C)C)c(c(c1)C(C)C)S(=O)(=O)n2cnc(n2)[N+]([O-])=O

InChI

1S/C17H24N4O4S/c1-10(2)13-7-14(11(3)4)16(15(8-13)12(5)6)26(24,25)20-9-18-17(19-20)21(22)23/h7-12H,1-6H3

InChI key

XJBNCASBZVSLGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Coupling reagent for oligonucleotide synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service