Skip to Content
Merck
All Photos(1)

Key Documents

243949

Sigma-Aldrich

Lithium aluminum hydride bis(tetrahydrofuran) solution

1.0 M in toluene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H20AlLiO2
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

concentration

1.0 M in toluene

density

0.875 g/mL at 25 °C

SMILES string

[Li+].[AlH4-].C1CCOC1.C2CCOC2

InChI

1S/2C4H8O.Al.Li.4H/c2*1-2-4-5-3-1;;;;;;/h2*1-4H2;;;;;;/q;;-1;+1;;;;

InChI key

VVFCZWUYZWVYGI-UHFFFAOYSA-N

Application

  • Reducing agent for preparation of of indazole- and benzisoxazole-isoindolines toward use in treatment of psychiatric disorders

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service