Skip to Content
Merck
All Photos(1)

Key Documents

243116

Sigma-Aldrich

2-sec-Butyl-3-methoxypyrazine

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€60.90

€60.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€60.90

About This Item

Empirical Formula (Hill Notation):
C9H14N2O
CAS Number:
Molecular Weight:
166.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€60.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

refractive index

n20/D 1.492 (lit.)

bp

50 °C/1 mmHg (lit.)

density

0.996 g/mL at 25 °C

SMILES string

CCC(C)c1nccnc1OC

InChI

1S/C9H14N2O/c1-4-7(2)8-9(12-3)11-6-5-10-8/h5-7H,4H2,1-3H3

InChI key

QMQDJVIJVPEQHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-sec-Butyl-3-methoxypyrazine was identified as the main source of earthy/bell pepper (EBP) flavor in the Farmstead Cheddar cheeses[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E R D Neta et al.
Journal of food science, 73(9), C632-C638 (2008-11-22)
Farmstead Cheddar cheeses with natural bandage wrappings have a distinctive flavor profile that is appealing to many consumers. An earthy/bell pepper (EBP) flavor has been previously recognized in some of these cheeses. This study characterized the alkylmethoxypyrazine compounds causing EBP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service