Skip to Content
Merck
All Photos(2)

Key Documents

242977

Sigma-Aldrich

Ethyl 2-aminobenzoate

≥99%

Synonym(s):

Ethyl anthranilate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€60.90

€60.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€60.90

About This Item

Linear Formula:
H2NC6H4CO2C2H5
CAS Number:
Molecular Weight:
165.19
Beilstein:
878874
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€60.90


Check Cart for Availability

Request a Bulk Order

vapor density

5.7 (vs air)

Assay

≥99%

form

liquid

refractive index

n20/D 1.564 (lit.)

bp

129-130 °C/9 mmHg (lit.)

mp

13-15 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1ccccc1N

InChI

1S/C9H11NO2/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6H,2,10H2,1H3

InChI key

TWLLPUMZVVGILS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 2-aminobenzoate (Ethyl anthranilate) is a minor constituent of Burgundy Pinot noir wines and is identified by coupled GC-MS analysis[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethyl anthranilate, ethyl cinnamate, 2, 3-dihydrocinnamate, and methyl anthranilate: four important odorants identified in Pinot noir wines of Burgundy.
Moio L and Etievant PX.
American Journal of Enology and Viticulture, 46(3), 392-398 (1995)
Guoying Sun et al.
Molecular vision, 17, 2191-2196 (2011-08-26)
Pirfenidone (5-methyl-1-phenyl-2-[1H]-pyridone) is a new, broad-spectrum agent that has an inhibition effect on the proliferation, migration, and collagen contraction of human Tenon's fibroblasts, and thus modulating the wound healing process of glaucoma filtering surgical site. This study investigated the pharmacokinetics
Tanya E Rutan et al.
Journal of agricultural and food chemistry, 66(38), 10053-10066 (2018-09-04)
The effects of two cluster thinning regimes (low and moderate) with Vitis vinifera cv. Pinot noir, in vineyards located in Central Otago, New Zealand, on wine composition, were studied across three consecutive seasons. There were strong correlations between the extent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service