Skip to Content
Merck
All Photos(1)

Key Documents

240869

Sigma-Aldrich

Styrene

ReagentPlus®, 99.9%

Synonym(s):

Phenylethylene, Vinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CH2
CAS Number:
Molecular Weight:
104.15
Beilstein:
1071236
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.6 (vs air)

vapor pressure

12.4 mmHg ( 37.7 °C)
4.3 mmHg ( 15 °C)

product line

ReagentPlus®

Assay

99.9%

form

liquid

autoignition temp.

914 °F

contains

10-15 ppm 4-tert-butylcatechol as inhibitor

expl. lim.

6.1 %

refractive index

n20/D 1.546 (lit.)

bp

145-146 °C (lit.)

mp

−31 °C (lit.)

density

0.906 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=Cc1ccccc1

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ping Qiu et al.
Frontiers in chemistry, 8, 130-130 (2020-03-27)
The halide perovskite (PVSK) material, an excellent light absorber with fast carrier kinetics, has received increased attention as a potential photocatalyst for organic synthesis. Herein, we report a straightforward synthesis of chemically modified halide perovskite and its application as an
Daniela Saviello et al.
Analytica chimica acta, 843, 59-72 (2014-08-26)
Synchrotron-based Fourier transform infrared micro-spectroscopy (SR-μFTIR) was used to map photo-oxidative degradation of acrylonitrile-butadiene-styrene (ABS) and to investigate the presence and the migration of additives in historical samples from important Italian design objects. High resolution (3×3 μm(2)) molecular maps were
A comprehensive evaluation of the potential health risks associated with occupational and environmental exposure to styrene.
Joshua T Cohen et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 5(1-2), 1-265 (2002-05-16)
Fanny Bonnet et al.
Dalton transactions (Cambridge, England : 2003), 44(27), 12312-12325 (2015-05-23)
New bis(phenolate)amine-supported neodymium borohydride complexes and their previously reported samarium analogues were tested as catalysts for the polymerisation of styrene and isoprene. Reaction of Na2O2N(L) (L = py, OMe, NMe2) with Nd(BH4)3(THF)3 afforded the borohydride complexes Nd(O2N(L))(BH4)(THF) (L = py
Visible light photocatalysis of [2+2] styrene cycloadditions by energy transfer.
Zhan Lu et al.
Angewandte Chemie (International ed. in English), 51(41), 10329-10332 (2012-09-12)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service