240346
Sulfanilamide
≥99%
Synonym(s):
p-Aminobenzenesulfonamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H2NC6H4SO2NH2
CAS Number:
Molecular Weight:
172.20
Beilstein:
511852
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
Assay
≥99%
mp
164-166 °C (lit.)
SMILES string
Nc1ccc(cc1)S(N)(=O)=O
InChI
1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)
InChI key
FDDDEECHVMSUSB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Edward E Knaus et al.
Bioorganic & medicinal chemistry letters, 21(19), 5892-5896 (2011-08-20)
A series of compounds incorporating regioisomeric phenylethynylbenzenesulfonamide moieties has been investigated for the inhibition of four human carbonic anhydrase (hCA, EC 4.2.1.1) isoforms, hCA I, II, IX and XII. Inhibition between the low nanomolar to the milliomolar range has been
Claudia Temperini et al.
Journal of medicinal chemistry, 53(2), 850-854 (2009-12-24)
Coumarins constitute a general and totally new class of inhibitors of the zinc enzyme carbonic anhydrase (CA, EC 4.2.1.1), binding at the entrance of the active site cavity. We report here that the coumarin-binding site in CAs may interact with
Daniela Vullo et al.
Bioorganic & medicinal chemistry letters, 20(7), 2178-2182 (2010-03-10)
A beta-carbonic anhydrase (CA, EC 4.2.1.1) from the bacterial pathogen Brucella suis, bsCA 1, has been cloned, purified characterized kinetically and for inhibition with a series of water soluble glycosylated sulfanilamides. bsCA 1 has appreciable activity as catalyst for the
Penghua Wang et al.
Water research, 45(16), 5015-5026 (2011-08-02)
A novel carbon-sensitized and nitrogen-doped TiO2 (C/N-TiO2) was synthesized by a facile sol-gel method using titanium butoxide as both titanium precursor and carbon source, and nitric acid as nitrogen source. The calcination temperature had a great effect on the crystal
Electrophilic trifluoromethanesulfanylation of organometallic species with trifluoromethanesulfanamides.
François Baert et al.
Angewandte Chemie (International ed. in English), 51(41), 10382-10385 (2012-09-15)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service